missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62894.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457966

  • 3114.20 SEK / 1g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD26143347

Specifications

4-Bromo-N-(4-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
Solid
97%
394.21
1g
MFCD26143347
Videos
SDS