missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide, 95%

Brand:  Alfa Aesar™ H65707.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479846

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide
398.22
MFCD28098814

Specifications

4-Bromo-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide
1g
398.22
Solid
95%
MFCD28098814
Videos
SDS