Läs mer
4-Bromo Riluzole, TRC
Product Code. 30136387
Change view
Click to view available options
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30136387 1 g
30007479 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136387 Supplier Toronto Research Chemicals Supplier No. TRCB6880701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144631-82-3
Molekylformel C8 H4 Br F3 N2 O S
Molekylvikt (g/mol) 313.09
Synonym 4-Bromo-6-(trifluoromethoxy)-2-benzothiazolamine, 4-Bromoriluzole
IUPAC-namn 4-bromo-6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
LEDER Nc1nc2c(Br)cc(OC(F)(F)F)cc2s1
Specifications
Kemiskt namn eller material 4-Bromo Riluzole
Kvantitet 1 g
InChI formel InChI=1S/C8H4BrF3N2OS/c9-4-1-3(15-8(10,11)12)2-5-6(4)14-7(13)16-5/h1-2H,(H2,13,14)
Formel vikt 311.918