missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromobenzaldehyde dimethyl acetal, 98%

Brand:  Alfa Aesar™ H55283.AA

Additional Details : CAS Number : 24856-58-4 Weight : 0.00005kg

Quantity 5mL

 View more versions of this product

Product Code. 15481488

  • 247.79 SEK / 5mL


Estimated Shipment date
from Supplier 01-03-2018
Add to basket

Description

Description

Applications
4-Bromobenzaldehyde dimethyl acetal, is used to produce other chemicals, it is used to produce 1-Bromo-4-bromomethyl-benzene at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

4-Bromobenzaldehyde dimethyl acetal
24856-58-4
231.09
MFCD01320499

Specifications

4-Bromobenzaldehyde dimethyl acetal
1.5320
254°C
Liquid
231.09
24856-58-4
1.383g/mL
5mL
98%
MFCD01320499
Videos
SDS