Learn More
4-Bromobenzenesulfinic acid sodium salt dihydrate, 97%, Thermo Scientific Chemicals
Catalog Number p-7024996
Click to view available options
Quantity:
2.5 g
10 g
50 g
175278-64-5
C6H4BrO2S
220.06
MFCD00210139
QBLQHDHWTVMHRH-UHFFFAOYSA-M
sodium 4-bromobenzenesulfinate dihydrate, 4-bromobenzenesulfinic acid sodium salt dihydrate, sodium 4-bromobenzene-1-sulphinate dihydrate, acmc-209e9v, c6h4bro2s.na.2h2o, sodium 4-bromophenylsulfinate dihydrate, sodium 4-bromobenzenesulfinate, dihydrate, sodium 4-bromanylbenzenesulfinate dihydrate, 4-bromobenzenesulfonic acid sodium salt 2-hydrate, sodium 4-bromobenzene-1-sulfinate-water 1/1/2
23683129
sodium;4-bromobenzenesulfinate;dihydrate
[O-]S(=O)C1=CC=C(Br)C=C1
175278-64-5
C6H4BrO2S
220.06
MFCD00210139
QBLQHDHWTVMHRH-UHFFFAOYSA-M
sodium 4-bromobenzenesulfinate dihydrate, 4-bromobenzenesulfinic acid sodium salt dihydrate, sodium 4-bromobenzene-1-sulphinate dihydrate, acmc-209e9v, c6h4bro2s.na.2h2o, sodium 4-bromophenylsulfinate dihydrate, sodium 4-bromobenzenesulfinate, dihydrate, sodium 4-bromanylbenzenesulfinate dihydrate, 4-bromobenzenesulfonic acid sodium salt 2-hydrate, sodium 4-bromobenzene-1-sulfinate-water 1/1/2
23683129
sodium;4-bromobenzenesulfinate;dihydrate
[O-]S(=O)C1=CC=C(Br)C=C1
4-Bromobenzenesulfinic acid sodium salt dehydrate is used in the preparation of unsymmetrical internal alkynes and vinyl sulfones by reaction with alkynes by using palladium as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobenzenesulfinic acid sodium salt dehydrate is used in the preparation of unsymmetrical internal alkynes and vinyl sulfones by reaction with alkynes by using palladium as a catalyst.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point >280°C
Quantity 2.5 g
Solubility Information Soluble in water.
Formula Weight 279.09 (243.06 Anhydrous)
Percent Purity 97%
Chemical Name or Material 4-Bromobenzenesulfinic acid sodium salt dihydrate