missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromobenzophenone, 98%

347.54 SEK - 2241.82 SEK

Chemical Identifiers

CAS 90-90-4
Molecular Formula C13H9BrO
Molecular Weight (g/mol) 261.118
MDL Number MFCD00000103
InChI Key KEOLYBMGRQYQTN-UHFFFAOYSA-N
Synonym 4-bromobenzophenone, 4-bromophenyl phenyl methanone, p-bromobenzophenone, methanone, 4-bromophenyl phenyl, benzophenone, 4-bromo, p-benzoylbromobenzene, usaf do-3, 4-bromophenyl phenyl ketone, 4-bromophenyl-phenyl-methanone, 4-bromophenyl-phenylmethanone
PubChem CID 7030
IUPAC Name (4-bromophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389409 SDS View Documents Alfa Aesar™
A14755.14
25g 347.54 SEK
25g View alternative products
Save Up to 
»
In stock, ready to ship: 3
Add to basket
 
11399409 SDS View Documents Alfa Aesar™
A14755.22
100g 1013.64 SEK
100g View alternative products
Save Up to 
»
Estimated Shipment
15-07-2020
Add to basket
 
11457806 SDS View Documents Alfa Aesar™
A14755.30
250g 2241.82 SEK
250g View alternative products
Save Up to 
»
Estimated Shipment
15-07-2020
Add to basket
 
Description

Description

Applications
4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

Solubility
Insoluble in water but slightly soluble in other solvents such as ethanol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

90-90-4
261.118
KEOLYBMGRQYQTN-UHFFFAOYSA-N
7030
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Br
C13H9BrO
MFCD00000103
4-bromobenzophenone, 4-bromophenyl phenyl methanone, p-bromobenzophenone, methanone, 4-bromophenyl phenyl, benzophenone, 4-bromo, p-benzoylbromobenzene, usaf do-3, 4-bromophenyl phenyl ketone, 4-bromophenyl-phenyl-methanone, 4-bromophenyl-phenylmethanone
(4-bromophenyl)-phenylmethanone

Specifications

4-Bromobenzophenone
350°C
25g
C13H9BrO
1910182
Insoluble in water but slightly soluble in other solvents such as ethanol,ether and benzene.
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Br
261.118
261.12
Odorless
79°C to 83°C
90-90-4
MFCD00000103
4-bromobenzophenone, 4-bromophenyl phenyl methanone, p-bromobenzophenone, methanone, 4-bromophenyl phenyl, benzophenone, 4-bromo, p-benzoylbromobenzene, usaf do-3, 4-bromophenyl phenyl ketone, 4-bromophenyl-phenyl-methanone, 4-bromophenyl-phenylmethanone
KEOLYBMGRQYQTN-UHFFFAOYSA-N
(4-bromophenyl)-phenylmethanone
7030
98%