missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylsulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7077365
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
53531-69-4
C7H6BrClO2S
269.55
ZWVWFWGJZPHCHF-UHFFFAOYSA-N
4-bromobenzylsulfonyl chloride, 4-bromophenyl methanesulfonyl chloride, 4-bromobenzylsulphonyl chloride, benzenemethanesulfonyl chloride, 4-bromo, 4-bromophenyl methanesulphonyl chloride, 4-bromophenyl methyl chlorosulfone, 4-bromo-phenyl-methanesulfonyl chloride, 4-bromo-benzenemethanesulfonyl chloride, pubchem2155, acmc-209l8j
598764
(4-bromophenyl)methanesulfonyl chloride
C1=CC(=CC=C1CS(=O)(=O)Cl)Br
This item is not returnable. View return policy
53531-69-4
C7H6BrClO2S
269.55
ZWVWFWGJZPHCHF-UHFFFAOYSA-N
4-bromobenzylsulfonyl chloride, 4-bromophenyl methanesulfonyl chloride, 4-bromobenzylsulphonyl chloride, benzenemethanesulfonyl chloride, 4-bromo, 4-bromophenyl methanesulphonyl chloride, 4-bromophenyl methyl chlorosulfone, 4-bromo-phenyl-methanesulfonyl chloride, 4-bromo-benzenemethanesulfonyl chloride, pubchem2155, acmc-209l8j
598764
(4-bromophenyl)methanesulfonyl chloride
C1=CC(=CC=C1CS(=O)(=O)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 116.0°C to 118.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 269.55
Percent Purity 95%
Chemical Name or Material 4-Bromobenzylsulfonyl chloride
compliance-icons
Product Identifier
  • 4-Bromobenzylsulfonyl chloride, 95%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'