missing translation for 'onlineSavingsMsg'
Learn More

4-Bromodiphenyl ether, 99%, Thermo Scientific™

Catalog Number p-4813080
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
101-55-3
C12H9BrO
249.11
JDUYPUMQALQRCN-UHFFFAOYSA-N
4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
7565
CHEBI:77421
1-bromo-4-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)Br
This item is not returnable. View return policy
101-55-3
C12H9BrO
249.11
JDUYPUMQALQRCN-UHFFFAOYSA-N
4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
7565
CHEBI:77421
1-bromo-4-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)Br
Melting Point 18.0°C
Density 1.4230g/mL
Boiling Point 305.0°C
Flash Point >113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OC6H5
Refractive Index 1.6081
Quantity 5 g
Beilstein 06,I,105
Specific Gravity 1.423
Formula Weight 249.11
Percent Purity 99%
Chemical Name or Material 4-Bromodiphenyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Bromodiphenyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'