Learn More
4-Bromomethyl-6,7-dimethoxycoumarin, 95%, Thermo Scientific Chemicals
Description
4-Bromomethyl-6,7-dimethoxycoumarin is used as fluorescent labeling reagent for the reversed-phase HPLC separation and detection of carboxylic acids. It is also used as derivatization reagent: for quantitation of clofibric, bezafibric and fenofibric acids in liver by liquid chromatography, in determination of the concentration of tripeptide glutathione by matrix-assisted laser desorption ionization time-of-flight mass spectrometry and for determination of carboxylic acids in chromatographic detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 200°C to 203°C |
Quantity | 0.5 g |
UN Number | UN3261 |
Beilstein | 4750487 |
Formula Weight | 299.12 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 4-Bromomethyl-6,7-dimethoxycoumarin |
RUO – Research Use Only