Learn More
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
Catalog Number p-6827172
Click to view available options
Quantity:
5 g
25 g
73870-24-3
C6H7Br2N
252.94
MFCD01863545
VAJUUDUWDNCECT-UHFFFAOYSA-N
4-bromomethyl pyridine hydrobromide, 4-bromomethylpyridine hydrobromide, 4-bromomethyl-pyridine hydrobromide, 4-pyridylmethyl bromide hydrobromide, pyridine, 4-bromomethyl-, hydrobromide, 4-bromomethyl pyridine, bromide, 4-bromomethyl-pyridine, 4-bromomethyl pyridinium bromide, acmc-1bbrw
12752219
4-(bromomethyl)pyridine;hydrobromide
[H+].[Br-].BrCC1=CC=NC=C1
73870-24-3
C6H7Br2N
252.94
MFCD01863545
VAJUUDUWDNCECT-UHFFFAOYSA-N
4-bromomethyl pyridine hydrobromide, 4-bromomethylpyridine hydrobromide, 4-bromomethyl-pyridine hydrobromide, 4-pyridylmethyl bromide hydrobromide, pyridine, 4-bromomethyl-, hydrobromide, 4-bromomethyl pyridine, bromide, 4-bromomethyl-pyridine, 4-bromomethyl pyridinium bromide, acmc-1bbrw
12752219
4-(bromomethyl)pyridine;hydrobromide
[H+].[Br-].BrCC1=CC=NC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 189.0°C to 192.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 252.93
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)pyridine hydrobromide
compliance-icons
Product Identifier
  • 4-(Bromomethyl)pyridine hydrobromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.