missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-2701628
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
5467-74-3
C6H6BBrO2
200.83
MFCD00002104
QBLFZIBJXUQVRF-UHFFFAOYSA-N
4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid
79599
(4-bromophenyl)boronic acid
OB(O)C1=CC=C(Br)C=C1
5467-74-3
C6H6BBrO2
200.83
MFCD00002104
QBLFZIBJXUQVRF-UHFFFAOYSA-N
4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid
79599
(4-bromophenyl)boronic acid
OB(O)C1=CC=C(Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 287.0°C to 292.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4B(OH)2
Quantity 5 g
Beilstein 16, II, 638
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,thf,dmso,dmf,methanol
Formula Weight 200.83
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'