Läs mer
4-Bromopyridazine Hydrobromide, TRC
Produktkod. 30348354
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30348354 500 mg
30335644 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30348354 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB686785500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Bromopyridazine Hydrobromide
CAS 1220039-64-4
Molekylformel C4 H3 Br N2 . Br H
Kvantitet 500 mg
InChI formel InChI=1S/C4H3BrN2.BrH/c5-4-1-2-6-7-3-4;/h1-3H;1H
LEDER Br.Brc1ccnnc1
IUPAC-namn 4-bromopyridazine;hydrobromide
Molekylvikt (g/mol) 239.9
Formel vikt 237.8741