Läs mer
4-Butoxy-2H-1-benzopyran-2-one, TRC
Product Code. 30343771
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30343771 1 g
1 options
Product Code. 30343771 Supplier Toronto Research Chemicals Supplier No. TRCB8098251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 4-Butoxy-2H-1-benzopyran-2-one
CAS 71386-90-8
Molekylformel C13H14O3
Kvantitet 1 g
Synonym 4-Butoxy Coumarin
InChI formel InChI=1S/C13H14O3/c1-2-3-8-15-12-9-13(14)16-11-7-5-4-6-10(11)12/h4-7,9H,2-3,8H2,1H3
LEDER CCCCOC(C1=CC=CC=C1O2)=CC2=O
IUPAC-namn 4-butoxychromen-2-one
Molekylvikt (g/mol) 218.25
Formel vikt 218.0943