missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Butylamino)benzoic Acid, Mikromol™
Description
4-(Butylamino)benzoic Acid
Specifications
Specifications
| Kemiskt namn eller material | 4-(Butylamino)benzoic Acid |
| CAS | 4740-24-3 |
| Analyt- eller komponentnamn | 4-(Butylamino)benzoic Acid |
| Frakt skick | Room Temperature |
| Molekylformel | C11 H15 N O2 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Tetracaine Hydrochloride Imp. B (EP), Tetracaine Imp. B (EP), Tetracaine USP Related Compound B, Tetracaine USP RC B, 4-(Butylamino)benzoic Acid, Tetracaine Related Compound B, Tetracaine Hydrochloride Impurity B, Tetracaine Impurity B |
| InChI formel | InChI=1S/C11H15NO2/c1-2-3-8-12-10-6-4-9(5-7-10)11(13)14/h4-7,12H,2-3,8H2,1H3,(H,13,14) |
| LEDER | CCCCNc1ccc(cc1)C(=O)O |
| IUPAC-namn | 4-(butylamino)benzoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?