Läs mer
4-(Butylamino)benzoic Acid, TRC
Produktkod. 30138368
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138368 5 g
30133753 10 g
17913935 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138368 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6894455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4740-24-3
Molekylformel C11 H15 N O2
Molekylvikt (g/mol) 193.24
Synonym 4-(Butylamino)benzoic acid, Tetracaine Hydrochloride Imp. B (EP), Benzoic acid, 4-(butylamino)-, Benzoic acid, p-(butylamino)- (6CI,7CI,8CI), NSC 44300, p-Butylaminobenzoic acid
IUPAC-namn 4-(butylamino)benzoic acid
LEDER CCCCNc1ccc(cc1)C(=O)O
Specifikationer
Kemiskt namn eller material 4-(Butylamino)benzoic Acid
Kvantitet 5 g
InChI formel InChI=1S/C11H15NO2/c1-2-3-8-12-10-6-4-9(5-7-10)11(13)14/h4-7,12H,2-3,8H2,1H3,(H,13,14)
Formel vikt 193.1103