missing translation for 'onlineSavingsMsg'
Learn More

4-Carbamoyltetrahydropyran, 97%, Acros Organics

Brand:  Acros Organics 450880050

Additional Details : CAS Number : 344329-76-6 Weight : 0.01000kg


Quantity 5g

 View more versions of this product

Product Code. 15239676

  • 1391.22 SEK / 5g


Estimated Shipment date
from Supplier 08-07-2019
Add to basket


Specifications

Chemical Identifiers

344329-76-6
129.159
tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide, 4-carbamoyltetrahydropyran, 4-carbamoyltetrahydro-2h-pyran, 2h-pyran-4-carboxamide, tetrahydro, 2h-3,4,5,6-tetrahydropyran-4-carboxamide, pubchem9735, acmc-209i7n, tetrahydro-2h-4-pyrancarboxamide, 2h-pyran-4-carboxamide,tetrahydro
oxane-4-carboxamide
C6H11NO2
DGOYLVBDCVINQZ-UHFFFAOYSA-N
13197203
C1COCCC1C(=O)N

Specifications

4-Carbamoyltetrahydropyran
5g
97%
tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide, 4-carbamoyltetrahydropyran, 4-carbamoyltetrahydro-2h-pyran, 2h-pyran-4-carboxamide, tetrahydro, 2h-3,4,5,6-tetrahydropyran-4-carboxamide, pubchem9735, acmc-209i7n, tetrahydro-2h-4-pyrancarboxamide, 2h-pyran-4-carboxamide,tetrahydro
C1COCCC1C(=O)N
129.159
129.16
180.0°C to 182.0°C
344329-76-6
C6H11NO2
DGOYLVBDCVINQZ-UHFFFAOYSA-N
oxane-4-carboxamide
13197203
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Carbamoyltetrahydropyran, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.