Learn More
4-Carbamoyltetrahydropyran, 97%, Thermo Scientific™
Catalog Number p-4813086
Click to view available options
Quantity:
1 g
5 g
344329-76-6
C6H11NO2
129.16
MFCD08235066
DGOYLVBDCVINQZ-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide, 4-carbamoyltetrahydropyran, 4-carbamoyltetrahydro-2h-pyran, 2h-pyran-4-carboxamide, tetrahydro, 2h-3,4,5,6-tetrahydropyran-4-carboxamide, pubchem9735, acmc-209i7n, tetrahydro-2h-4-pyrancarboxamide, 2h-pyran-4-carboxamide,tetrahydro
13197203
oxane-4-carboxamide
NC(=O)C1CCOCC1
344329-76-6
C6H11NO2
129.16
MFCD08235066
DGOYLVBDCVINQZ-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide, 4-carbamoyltetrahydropyran, 4-carbamoyltetrahydro-2h-pyran, 2h-pyran-4-carboxamide, tetrahydro, 2h-3,4,5,6-tetrahydropyran-4-carboxamide, pubchem9735, acmc-209i7n, tetrahydro-2h-4-pyrancarboxamide, 2h-pyran-4-carboxamide,tetrahydro
13197203
oxane-4-carboxamide
NC(=O)C1CCOCC1
Melting Point 180.0°C to 182.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.16
Percent Purity 97%
Chemical Name or Material 4-Carbamoyltetrahydropyran
compliance-icons
Product Identifier
  • 4-Carbamoyltetrahydropyran, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.