Läs mer
4-[[(Carbomethoxy)methyl]amino]phenol, TRC
Produktkod. 30350558
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30350558 500 mg
1 options
Produktkod. 30350558 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC564050500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(((Carbomethoxy)methyl)amino)phenol
CAS 56405-21-1
Molekylformel C9 H11 N O3
Kvantitet 500 mg
Synonym 4-[[(Carbomethoxy)methyl]amino]phenol, N-(4-Hydroxyphenyl)glycine Methyl Ester, (4-Hydroxy-phenylamino)-acetic Acid Methyl Ester, NSC 145152, [N-(4-Hydroxyphenyl)amino]acetic Acid Methyl Ester
InChI formel InChI=1S/C9H11NO3/c1-13-9(12)6-10-7-2-4-8(11)5-3-7/h2-5,10-11H,6H2,1H3
LEDER COC(=O)CNc1ccc(O)cc1
IUPAC-namn methyl 2-(4-hydroxyanilino)acetate
Molekylvikt (g/mol) 181.189
Formel vikt 181.074