Läs mer
(4-Carboxy-3-ethoxy)phenyl Acetic Acid, TRC
Produktkod. 30367389
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367389 5 mg
30353871 25 mg
30365996 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30367389 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1779605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 220438-80-2
Molekylformel C11 H12 O5
Molekylvikt (g/mol) 224.21
Synonym Benzeneacetic acid, 4-carboxy-3-ethoxy-, 4-Carboxy-3-ethoxybenzeneacetic acid, 4-Carboxymethyl-2-ethoxybenzoic acid, Repaglinide Imp. A (EP)
IUPAC-namn 4-(carboxymethyl)-2-ethoxybenzoic acid
LEDER CCOc1cc(CC(=O)O)ccc1C(=O)O
Specifikationer
Kemiskt namn eller material (4-Carboxy-3-ethoxy)phenyl Acetic Acid
Kvantitet 5 mg
InChI formel InChI=1S/C11H12O5/c1-2-16-9-5-7(6-10(12)13)3-4-8(9)11(14)15/h3-5H,2,6H2,1H3,(H,12,13)(H,14,15)
Formel vikt 224.0685