Läs mer
4-Carboxy-a-methylbenzeneacetic Acid, TRC
Produktkod. 30364372
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30364372 25 mg
1 options
Produktkod. 30364372 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17875525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Carboxy-alpha-methylbenzeneacetic Acid
CAS 67381-50-4
Molekylformel C10H10O4
Kvantitet 25 mg
Synonym Benzeneacetic acid, 4-carboxy-α-methyl-, 4-Carboxy-α-methylbenzeneacetic acid, 2-p-Carboxyphenylpropionic acid, 4-(1-Carboxyethyl)benzoic acid, DL-2-(4-Carboxyphenyl)propionic acid, Carboxyhydratropic acid (CA-HA)
InChI formel InChI=1S/C10H10O4/c1-6(9(11)12)7-2-4-8(5-3-7)10(13)14/h2-6H,1H3,(H,11,12)(H,13,14)
LEDER CC(C(=O)O)c1ccc(cc1)C(=O)O
IUPAC-namn 4-(1-carboxyethyl)benzoic acid
Molekylvikt (g/mol) 194.18
Formel vikt 194.0579