Läs mer
4-Carboxy Nevirapine, TRC
Produktkod. 30368184
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368184 50 mg
30359726 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18105050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 245501-02-4
Molekylformel C15H12N4O3
Molekylvikt (g/mol) 296.28
Synonym 11-Cyclopropyl-6,11-dihydro-6-oxo-5H-dipyrido[3,2-b:2',3'-e][1,4]diazepine-4-carboxylic Acid
IUPAC-namn 11-cyclopropyl-6-oxo-5H-dipyrido[3,2-a:2',3'-d][1,4]diazepine-4-carboxylic acid
LEDER OC(=O)c1ccnc2N(C3CC3)c4ncccc4C(=O)Nc12
Specifikationer
Kemiskt namn eller material 4-Carboxy Nevirapine
Kvantitet 50 mg
InChI formel InChI=1S/C15H12N4O3/c20-14-10-2-1-6-16-12(10)19(8-3-4-8)13-11(18-14)9(15(21)22)5-7-17-13/h1-2,5-8H,3-4H2,(H,18,20)(H,21,22)
Formel vikt 296.0909