Läs mer
4-Carboxymethylphenylboronic acid, TRC
Produktkod. 30352150
Change view
Kvantitet:
50 mg
Product Code. Kvantitet
30352150 50 mg
1 options
Product Code. 30352150 Supplier Toronto Research Chemicals Supplier No. TRCC18104850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 4-Carboxymethylphenylboronic acid
CAS 90111-58-3
Molekylformel C8H9BO4
Kvantitet 50 mg
InChI formel InChI=1S/C8H9BO4/c10-8(11)5-6-1-3-7(4-2-6)9(12)13/h1-4,12-13H,5H2,(H,10,11)
LEDER OB(O)c1ccc(CC(=O)O)cc1
IUPAC-namn 2-(4-boronophenyl)acetic acid
Molekylvikt (g/mol) 179.97
Formel vikt 180.0594