Learn More
4'-Chloro-[1,1'-biphenyl]-4-ol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 28034-99-3 |
| Molecular Formula | C12H9ClO |
| MDL Number | MFCD00143298 |
| Quantity | 5 g |
| SMILES | OC1=CC=C(C2=CC=C(Cl)C=C2)C=C1 |
| Molecular Weight (g/mol) | 204.65 |
| Percent Purity | 98% |
| Chemical Name or Material | 4'-Chloro-[1,1'-biphenyl]-4-ol |