Läs mer
4-Chloro-1,1’-biphenyl, TRC
Produktkod. 30357763
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357763 100 mg
30350449 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30357763 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC365053100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2051-62-9
Molekylformel C12 H9 Cl
Molekylvikt (g/mol) 188.65
Synonym 1,1'-Biphenyl, 4-chloro-, Biphenyl, 4-chloro- (6CI,7CI,8CI), 4-Chloro-1,1'-biphenyl, 1,1'-Biphenyl-4-yl chloride, 1-Chloro-4-phenylbenzene, 4-Biphenylyl chloride, 4-Chlorobiphenyl, 4-Chlorodiphenyl, NSC 37066, PCB 3, p-Chlorobiphenyl, 4-MonoCB
IUPAC-namn 1-chloro-4-phenylbenzene
LEDER Clc1ccc(cc1)c2ccccc2
Specifikationer
Kemiskt namn eller material PCB 3 (4-Chlorobiphenyl)
Kvantitet 100 mg
InChI formel InChI=1S/C12H9Cl/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H
Formel vikt 188.0393