Läs mer
4-Chloro-1-methylpiperidine, TRC
Produktkod. 30350351
Change view
Kvantitet:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350351 50 g
30363254 25 g
30368088 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30350351 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37107050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5570-77-4
Molekylformel C6H12ClN
Molekylvikt (g/mol) 133.62
Synonym 1-Methyl-4-chloropiperidine, 1-Methyl-4-piperidinyl Chloride, 1-Methyl-4-piperidyl Chloride, 4-Chloro-N-methylpiperidine, N-Methyl-4-chloropiperidine
IUPAC-namn 4-chloro-1-methylpiperidine
LEDER CN1CCC(Cl)CC1
Specifikationer
Kemiskt namn eller material 4-Chloro-1-methylpiperidine
Kvantitet 50 g
InChI formel InChI=1S/C6H12ClN/c1-8-4-2-6(7)3-5-8/h6H,2-5H2,1H3