Läs mer
4-Chloro-2,3-dimethylpyridine N-Oxide, Mikromol™
Produktkod. 30049424
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049424 25 mg
17952526 100 mg
30462518 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30049424 Leverantör Mikromol Leverantörsnummer MM0618.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Chloro-2,3-dimethylpyridine N-Oxide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Chloro-2,3-dimethylpyridine N-Oxide
CAS 59886-90-7
Analyt- eller komponentnamn 4-Chloro-2,3-dimethylpyridine N-Oxide
Frakt skick Room Temperature
Molekylformel C7 H8 Cl N O
Certifikat och efterlevnad ISO 9001
Synonym 4-Chloro-2,3-dimethylpyridine N-Oxide
InChI formel InChI=1S/C7H8ClNO/c1-5-6(2)9(10)4-3-7(5)8/h3-4H,1-2H3
LEDER Cc1c(C)[n+]([O-])ccc1Cl
IUPAC-namn 4-chloro-2,3-dimethyl-1-oxidopyridin-1-ium
Molekylvikt (g/mol) 157.6
Kvantitet 25 mg
Formel vikt 157.0294 g/mol