missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2'-fluoro-3'-methoxyacetophenone, 97%, Thermo Scientific™

Catalog Number p-7052511
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-44-4
C9H8ClFO2
202.61
MFCD19687210
SSQUECJMKSQDHG-UHFFFAOYSA-N
4'-chloro-2'-fluoro-3'-methoxyacetophenone, 1-4-chloro-2-fluoro-3-methoxyphenyl ethanone
66523544
1-(4-chloro-2-fluoro-3-methoxyphenyl)ethanone
COC1=C(Cl)C=CC(C(C)=O)=C1F
This item is not returnable. View return policy
1323966-44-4
C9H8ClFO2
202.61
MFCD19687210
SSQUECJMKSQDHG-UHFFFAOYSA-N
4'-chloro-2'-fluoro-3'-methoxyacetophenone, 1-4-chloro-2-fluoro-3-methoxyphenyl ethanone
66523544
1-(4-chloro-2-fluoro-3-methoxyphenyl)ethanone
COC1=C(Cl)C=CC(C(C)=O)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 202.61
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4'-Chloro-2'-fluoro-3'-methoxyacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'