missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorophenol, 98%, Thermo Scientific™

Catalog Number 15497948
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
348-62-9
C6H4ClFO
146.55
MFCD00075030
ZKMUKBBWORLNLA-UHFFFAOYSA-N
2-fluoro-4-chlorophenol, phenol, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenol, 4-chloro-fluorophenol, pubchem2861, acmc-1ct1y, ksc222g4r, 4-chloro-2-fluorophenol, attercop-chm at106713
181975
4-chloro-2-fluorophenol
OC1=CC=C(Cl)C=C1F
This item is not returnable. View return policy
348-62-9
C6H4ClFO
146.55
MFCD00075030
ZKMUKBBWORLNLA-UHFFFAOYSA-N
2-fluoro-4-chlorophenol, phenol, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenol, 4-chloro-fluorophenol, pubchem2861, acmc-1ct1y, ksc222g4r, 4-chloro-2-fluorophenol, attercop-chm at106713
181975
4-chloro-2-fluorophenol
OC1=CC=C(Cl)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 20°C
Color Colorless to Yellow
Density 1.378 g/mL
Boiling Point 103°C to 104°C
Flash Point 81°C (178°F)
Refractive Index 1.536
Quantity 5 g
Formula Weight 146.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-fluorophenol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'