missing translation for 'onlineSavingsMsg'
Läs mer

4-Chloro-2-formylbenzoic Acid, TRC

Produktkod. 30368290
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30368290 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368290 Supplier Toronto Research Chemicals Supplier No. TRCC371280100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Chloro-2-formylbenzoic Acid
CAS 4657-56-1
Molekylformel C8H5ClO3
Kvantitet 100 mg
Synonym 4-Chloro-phthalaldehydic Acid, 4-Chloro-2-formylbenzoic Acid
InChI formel InChI=1S/C8H5ClO3/c9-6-1-2-7(8(11)12)5(3-6)4-10/h1-4H,(H,11,12)
LEDER ClC1=CC(C([H])=O)=C(C(O)=O)C=C1
IUPAC-namn 4-chloro-2-formylbenzoic acid
Molekylvikt (g/mol) 184.58
Formel vikt 183.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.