Läs mer
4-Chloro-2-methyl-1-pentene, TRC
Produktkod. 30366989
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366989 50 mg
30364074 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366989 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36402550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 344322-55-0
Molekylformel C6H11Cl
Molekylvikt (g/mol) 118.6
IUPAC-namn 4-chloro-2-methylpent-1-ene
LEDER CC(Cl)CC(=C)C
Specifikationer
Kemiskt namn eller material 4-Chloro-2-methyl-1-pentene
Kvantitet 50 mg
InChI formel InChI=1S/C6H11Cl/c1-5(2)4-6(3)7/h6H,1,4H2,2-3H3
Formel vikt 118.0549