Läs mer
4-Chloro-2-nitrodiphenylamine, TRC
Produktkod. 30362587
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30362587 100 mg
1 options
Produktkod. 30362587 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC382525100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Disperse Yellow 26
CAS 16611-15-7
Molekylformel C12 H9 Cl N2 O2
Kvantitet 100 mg
Synonym Diphenylamine, 4-chloro-2-nitro- (6CI,8CI), 4-Chloro-2-nitro-N-phenylbenzenamine, 4-Chloro-2-nitrodiphenylamine, C.I. 10348, C.I. Disperse Yellow 26, Disperse Yellow 26, NSC 403663
InChI formel InChI=1S/C12H9ClN2O2/c13-9-6-7-11(12(8-9)15(16)17)14-10-4-2-1-3-5-10/h1-8,14H
LEDER [O-][N+](=O)c1cc(Cl)ccc1Nc2ccccc2
IUPAC-namn 4-chloro-2-nitro-N-phenylaniline
Molekylvikt (g/mol) 248.67
Formel vikt 248.0353