missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)phenylboronic acid, 95%, Thermo Scientific™

Catalog Number p-4602040
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
313545-41-4
C7H5BClF3O2
224.38
YAPBOBGBYQQYHX-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethylphenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl-phenylboronic acid, boronic acid, 4-chloro-2-trifluoromethyl phenyl, 4-chloro-2-trifluoromethyl phenylboronicacid, pubchem5158, acmc-20aic7
2763244
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
This item is not returnable. View return policy
313545-41-4
C7H5BClF3O2
224.38
YAPBOBGBYQQYHX-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethylphenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl-phenylboronic acid, boronic acid, 4-chloro-2-trifluoromethyl phenyl, 4-chloro-2-trifluoromethyl phenylboronicacid, pubchem5158, acmc-20aic7
2763244
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 224.38
Percent Purity 95%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)phenylboronic acid
compliance-icons
Product Identifier
  • 4-Chloro-2-(trifluoromethyl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'