missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluoropyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-4813093
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2546-56-7
C5H3ClFN
131.53
MFCD03453233
BEQUUSCRAKEKQM-UHFFFAOYSA-N
pyridine, 4-chloro-3-fluoro, 3-fluoro-4-chloro pyridine, 3-fluoro-4-chloropyridine hydrochloride, pubchem6667, 4-chloro-3fluoropyridine, 3-fluoro-4-chloropyridine, acmc-209x6p, 4-chloro-3-fluoro-pyridin, 3-fluoro-4-chloro-pyridine, 4-chloro-3-fluoro-pyridine
3852429
4-chloro-3-fluoropyridine
FC1=C(Cl)C=CN=C1
This item is not returnable. View return policy
2546-56-7
C5H3ClFN
131.53
MFCD03453233
BEQUUSCRAKEKQM-UHFFFAOYSA-N
pyridine, 4-chloro-3-fluoro, 3-fluoro-4-chloro pyridine, 3-fluoro-4-chloropyridine hydrochloride, pubchem6667, 4-chloro-3fluoropyridine, 3-fluoro-4-chloropyridine, acmc-209x6p, 4-chloro-3-fluoro-pyridin, 3-fluoro-4-chloro-pyridine, 4-chloro-3-fluoro-pyridine
3852429
4-chloro-3-fluoropyridine
FC1=C(Cl)C=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -27°C to 24°C
Boiling Point 142°C to 143°C
Flash Point 28°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 131.54
Percent Purity 95%
Chemical Name or Material 4-Chloro-3-fluoropyridine
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-3-fluoropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'