missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-iodobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7077394
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
42860-04-8
C7H4ClIO2
282.46
MFCD00079732
SDRURVZKYHGDAP-UHFFFAOYSA-N
3-iodo-4-chlorobenzoic acid, 4-chloro-3-iodo-benzoic acid, benzoic acid, 4-chloro-3-iodo, pubchem12500, 4-chloro-3-iodo benzoicacid, benzoic acid,4-chloro-3-iodo, zerenex e/9071913, buttpark 100\01-50
2757616
4-chloro-3-iodobenzoic acid
OC(=O)C1=CC(I)=C(Cl)C=C1
This item is not returnable. View return policy
42860-04-8
C7H4ClIO2
282.46
MFCD00079732
SDRURVZKYHGDAP-UHFFFAOYSA-N
3-iodo-4-chlorobenzoic acid, 4-chloro-3-iodo-benzoic acid, benzoic acid, 4-chloro-3-iodo, pubchem12500, 4-chloro-3-iodo benzoicacid, benzoic acid,4-chloro-3-iodo, zerenex e/9071913, buttpark 100\01-50
2757616
4-chloro-3-iodobenzoic acid
OC(=O)C1=CC(I)=C(Cl)C=C1
Melting Point 218.0°C to 219.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 282.47
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-iodobenzoic acid
compliance-icons
Product Identifier
  • 4-Chloro-3-iodobenzoic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'