missing translation for 'onlineSavingsMsg'
Läs mer

4-Chloro-3-nitro-5-sulfamoylbenzoic Acid, TRC

Produktkod. 30360023
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360023 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30360023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3695801G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 4-Chloro-3-nitro-5-sulfamoylbenzoic Acid
CAS 22892-96-2
Molekylformel C7 H5 Cl N2 O6 S
Kvantitet 1 g
InChI formel InChI=1S/C7H5ClN2O6S/c8-6-4(10(13)14)1-3(7(11)12)2-5(6)17(9,15)16/h1-2H,(H,11,12)(H2,9,15,16)
LEDER NS(=O)(=O)c1cc(cc(c1Cl)[N+](=O)[O-])C(=O)O
IUPAC-namn 4-chloro-3-nitro-5-sulfamoylbenzoic acid
Molekylvikt (g/mol) 280.64
Formel vikt 279.9557
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.