Learn More
Alfa Aesar™ 4-Chloro-3-sulfamoyl-N-[4-(trifluoromethyl)phenyl]benzamide, 95%
Brand:  Alfa Aesar™ H65651.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449806
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 12-11-2018
Add to basket
Specifications
Chemical Identifiers
MFCD27975798
Specifications
4-Chloro-3-sulfamoyl-N-[4-(trifluoromethyl)phenyl]benzamide
MFCD27975798
Solid
1g
378.75
95%