Learn More
4-Chloro-3-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Chloro-3-(trifluoromethoxy)benzyl bromide is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
4-Chloro-3-(trifluoromethoxy)benzyl bromide is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Specifications
Quantity | 250 mg |
UN Number | UN3265 |
Solubility Information | Not miscible or difficult to mix with water. |
Formula Weight | 289.48 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloro-3-(trifluoromethoxy)benzyl bromide |