Läs mer
4-Chloro-4'-hydroxybiphenyl, TRC
Produktkod. 30354393
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354393 100 mg
30362860 50 mg
30360400 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354393 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC585835100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28034-99-3
Molekylformel C12H9OCl
Molekylvikt (g/mol) 204.65
Synonym 4-Hydroxy-4'-chlorobiphenyl, 4'-Chloro-4-biphenylol, 4'-Hydroxy-4-chlorobiphenyl, p-Chloro-p'-hydroxybiphenyl, 4'-Chloro[1,1'-biphenyl]-4-ol, 4-(4-Chlorophenyl)phenol, 4-(4'-Chlorophenyl)phenol, 4-Chloro-4'-biphenylol
IUPAC-namn 4-(4-chlorophenyl)phenol
LEDER Oc1ccc(cc1)c2ccc(Cl)cc2
Specifikationer
Kemiskt namn eller material 4-Chloro-4'-hydroxybiphenyl
Kvantitet 100 mg
InChI formel InChI=1S/C12H9ClO/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,14H
Formel vikt 204.0342