Läs mer
4-Chloro-5-sulfamoyl-2-[[((2RS)-tetrahydrofuran-2-yl)methyl]amino]benzoic Acid, Mikromol™
Produktkod. 30459514
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30459514 4 x 25 mg
30051284 100 mg
30049408 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30459514 Leverantör Mikromol Leverantörsnummer MM0014.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Chloro-5-sulfamoyl-2-[[((2RS)-tetrahydrofuran-2-yl)methyl]amino]benzoic Acid, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetrahydro Furosemide
CAS 4793-38-8
Analyt- eller komponentnamn Tetrahydro Furosemide
Frakt skick Room Temperature
Molekylformel C12 H15 Cl N2 O5 S
Certifikat och efterlevnad ISO 17025
Synonym 5-(Aminosulfonyl)-4-chloro-2-[[(tetrahydro-2-furanyl)methyl]amino]benzoic acid, Anthranilic acid, 4-chloro-5-sulfamoyl-N-(tetrahydrofurfuryl)- (7CI,8CI), Furosemide Imp. F (EP), Furosemide Impurity F
InChI formel InChI=1S/C12H15ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h4-5,7,15H,1-3,6H2,(H,16,17)(H2,14,18,19)
Lösningstyp Neat
LEDER NS(=O)(=O)c1cc(C(=O)O)c(NCC2CCCO2)cc1Cl
IUPAC-namn 4-chloro-2-(oxolan-2-ylmethylamino)-5-sulfamoylbenzoic acid
Molekylvikt (g/mol) 334.78
Kvantitet 4 x 25 mg
Formel vikt 334.039 g/mol