Läs mer
4-Chloro-5-sulfamoylanthranilic Acid Methyl Ester, TRC
Produktkod. 30353505
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30353505 25 mg
1 options
Produktkod. 30353505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37512525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Chloro-5-sulfamoylanthranilic Acid Methyl Ester
CAS 3994-89-6
Molekylformel C8H9ClN2O4S
Kvantitet 25 mg
Synonym 2-Amino-5-(aminosulfonyl)-4-chlorobenzoic Acid Methyl Ester
InChI formel InChI=1S/C8H9ClN2O4S/c1-15-8(12)4-2-7(16(11,13)14)5(9)3-6(4)10/h2-3H,10H2,1H3,(H2,11,13,14)
LEDER COC(=O)c1cc(c(Cl)cc1N)S(=O)(=O)N
IUPAC-namn methyl 2-amino-4-chloro-5-sulfamoylbenzoate
Molekylvikt (g/mol) 264.69
Formel vikt 263.9972