Läs mer
4-Chloro-6,7-dimethoxyquinoline, TRC
Produktkod. 30012581
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012581 5 g
30005149 1 g
30011623 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012581 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3659305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35654-56-9
Molekylformel C11H10ClNO2
Molekylvikt (g/mol) 223.66
Synonym Quinoline, 4-chloro-6,7-dimethoxy- (9CI, ACI), 4-Chloro-6,7-dimethoxyquinoline (ACI)
IUPAC-namn 4-chloro-6,7-dimethoxyquinoline
LEDER COc1cc2nccc(Cl)c2cc1OC
Specifikationer
Kemiskt namn eller material 4-Chloro-6,7-dimethoxyquinoline
Kvantitet 5 g
InChI formel InChI=1S/C11H10ClNO2/c1-14-10-5-7-8(12)3-4-13-9(7)6-11(10)15-2/h3-6H,1-2H3
Formel vikt 223.04