Läs mer
4-Chloro-6-(trifluoromethyl)-3-pyridinecarboxylic Acid, TRC
Produktkod. 30364089
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364089 100 mg
30360920 500 mg
30352595 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC253066100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1060810-66-3
Molekylformel C7H3NO2F3Cl
Molekylvikt (g/mol) 225.55
IUPAC-namn 4-chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
LEDER OC(=O)c1cnc(cc1Cl)C(F)(F)F
Specifikationer
Kemiskt namn eller material 4-Chloro-6-(trifluoromethyl)-3-pyridinecarboxylic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C7H3ClF3NO2/c8-4-1-5(7(9,10)11)12-2-3(4)6(13)14/h1-2H,(H,13,14)
Formel vikt 224.9804