Läs mer
4-Chloro-benzoic Acid 1-(4-Methoxyphenyl)hydrazide Hydrochloride, TRC
Produktkod. 30352742
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30352742 25 mg
1 options
Produktkod. 30352742 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36443025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Chloro-benzoic Acid 1-(4-Methoxyphenyl)hydrazide Hydrochloride
CAS 13815-62-8
Molekylformel C14 H13 Cl N2 O2 . H Cl
Kvantitet 25 mg
Synonym 4-Chloro-benzoic Acid 1-(4-Methoxyphenyl)hydrazide Monohydrochloride, p-Chloro-benzoic Acid 1-(p-Methoxyphenyl)hydrazide Monohydrochloride
InChI formel InChI=1S/C14H13ClN2O2.ClH/c1-19-13-8-6-12(7-9-13)17(16)14(18)10-2-4-11(15)5-3-10;/h2-9H,16H2,1H3;1H
LEDER Cl.COc1ccc(cc1)N(N)C(=O)c2ccc(Cl)cc2
IUPAC-namn 4-chloro-N-(4-methoxyphenyl)benzohydrazide;hydrochloride
Molekylvikt (g/mol) 313.18
Formel vikt 312.0432