Läs mer
4-Chloro Kynurenine, TRC
Produktkod. 30138137
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138137 10 mg
30138637 100 mg
30011688 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138137 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36694010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75802-84-5
Molekylformel C10H11ClN2O3
Molekylvikt (g/mol) 242.66
Synonym α,2-Diamino-4-chloro-γ-oxobenzenebutanoic Acid, (+/-)-α,2-Diamino-4-chloro-γ-oxobenzenebutanoic Acid, 4-Cl-KYN, (+/-)-AV-101;
IUPAC-namn 2-amino-4-(2-amino-4-chlorophenyl)-4-oxobutanoic acid
LEDER C1=CC(=C(C=C1Cl)N)C(=O)CC(C(=O)O)N
Specifikationer
Kemiskt namn eller material 4-Chloro Kynurenine
Kvantitet 10 mg
InChI formel InChI=1S/C10H11ClN2O3/c11-5-1-2-6(7(12)3-5)9(14)4-8(13)10(15)16/h1-3,8H,4,12-13H2,(H,15,16)
Formel vikt 242.0458