Läs mer
4-Chloro-L-phenylalanine Hydrochloride, TRC
Produktkod. 30178960
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30178960 250 mg
30178983 100 mg
30178990 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30178960 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC382308250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 123053-23-6
Molekylformel C9H10ClNO2 . HCl
Molekylvikt (g/mol) 236.09
Synonym (L)-4-Chlorophenylalanine Hydrochloride;
IUPAC-namn (2S)-2-amino-3-(4-chlorophenyl)propanoic acid;hydrochloride
LEDER Cl.N[C@@H](Cc1ccc(Cl)cc1)C(=O)O
Specifikationer
Kemiskt namn eller material 4-Chloro-L-phenylalanine Hydrochloride
Kvantitet 250 mg
InChI formel InChI=1S/C9H10ClNO2.ClH/c10-7-3-1-6(2-4-7)5-8(11)9(12)13;/h1-4,8H,5,11H2,(H,12,13);1H/t8-;/m0./s1