Learn More
Alfa Aesar™ 4-Chloro-N-(2,4-dichlorophenyl)-2-fluoro-5-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65523.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15489706
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 06-02-2018
Add to basket
Specifications
Chemical Identifiers
4-Chloro-N-(2,4-dichlorophenyl)-2-fluoro-5-sulfamoylbenzamide
397.64
Specifications
4-Chloro-N-(2,4-dichlorophenyl)-2-fluoro-5-sulfamoylbenzamide
Solid
397.64
1g
95%