Learn More
Alfa Aesar™ 4-Chloro-N-(2-chloro-4-methylphenyl)-3-sulfamoylbenzamide, 96%
Brand:  Alfa Aesar™ H65435.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15489636
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
4-Chloro-N-(2-chloro-4-methylphenyl)-3-sulfamoylbenzamide
359.23
Specifications
4-Chloro-N-(2-chloro-4-methylphenyl)-3-sulfamoylbenzamide
Off-white
96%
1g
Solid
359.23