missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3,5-dimethoxyphenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65940.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450027

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3,5-dimethoxyphenyl)-3-sulfamoylbenzamide
370.81
MFCD27975842

Specifications

4-Chloro-N-(3,5-dimethoxyphenyl)-3-sulfamoylbenzamide
Solid
370.81
1g
95%
MFCD27975842
Videos
SDS