missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-chlorophenyl)-2-fluoro-5-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65225.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499476

  • 2716.64 SEK / 1g


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD28098772

Specifications

4-Chloro-N-(3-chlorophenyl)-2-fluoro-5-sulfamoylbenzamide
MFCD28098772
Solid
1g
363.19
95%
Videos
SDS