Learn More
Alfa Aesar™ 4-Chloro-N-(4-chloro-3-methylphenyl)-3-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65561.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15489736
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
4-Chloro-N-(4-chloro-3-methylphenyl)-3-sulfamoylbenzamide
359.23
MFCD27975786
Specifications
4-Chloro-N-(4-chloro-3-methylphenyl)-3-sulfamoylbenzamide
Solid
359.23
1g
95%
MFCD27975786